Learn More
Meso Scale Discovery UPLEX HUM RANTES ASYQP 25 PL
SDP
Catalog No. 5000079435
Encompass
The Meso Scale Discovery U-PLEX Human RANTES Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of RANTES (CCL5) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology providing high sensitivity and a wide dynamic range The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a lower limit of detection of 0 06 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-79435 Supplier Meso Scale Discovery Supplier No. K151A2K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RANTES Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of RANTES (CCL5) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology providing high sensitivity and a wide dynamic range The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a lower limit of detection of 0 06 pg/mL and an upper limit of 10 000 pg/mL