Learn More
Meso Scale Discovery UPLEX HUM RBP4 ASY384well
SDP
Catalog No. 5000081127
Encompass
The Meso Scale Discovery U-PLEX Human RBP4 384-Well Assay is a multiplex immunoassay kit for the quantitative measurement of Retinol Binding Protein 4 (RBP4) in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples The assay has a detection range of 50-10 000 pg/mL and includes reagents for standard curve generation The total assay time is 3 5 hours This product can aid in the research of RBP4 and its role in conditions like obesity and insulin resistance
Catalog No. 50-000-81127 Supplier Meso Scale Discovery Supplier No. K251V9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RBP4 384-Well Assay is a multiplex immunoassay kit for the quantitative measurement of Retinol Binding Protein 4 (RBP4) in human serum plasma and cell culture supernatants The assay utilizes MSD s electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples The assay has a detection range of 50-10 000 pg/mL and includes reagents for standard curve generation The total assay time is 3 5 hours This product can aid in the research of RBP4 and its role in conditions like obesity and insulin resistance