Learn More
Meso Scale Discovery UPLEX HUM RBP4 ASYQP 25 PL
SDP
Catalog No. 5000080401
Encompass
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible detection of RBP4 levels with high specificity and low background The assay requires a small sample volume and provides a rapid 3 5 hour protocol The U-PLEX platform provides researchers with an efficient tool for studying RBP4 biology and its role in conditions like insulin resistance diabetes and obesity
Catalog No. 50-000-80401 Supplier Meso Scale Discovery Supplier No. K151V9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human RBP4 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of retinol binding protein 4 (RBP4) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible detection of RBP4 levels with high specificity and low background The assay requires a small sample volume and provides a rapid 3 5 hour protocol The U-PLEX platform provides researchers with an efficient tool for studying RBP4 biology and its role in conditions like insulin resistance diabetes and obesity