Learn More
Meso Scale Discovery UPLEX HUM S100A12 ASY384well
SDP
Catalog No. 5000081121
Encompass
The Meso Scale Discovery U-PLEX Human S100A12 Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of S100A12 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires a small sample volume and has a testing time of approximately 3 5 hours It includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls The kit can perform 38 samples in duplicate along with calibrators and controls
Catalog No. 50-000-81121 Supplier Meso Scale Discovery Supplier No. K251AHPK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human S100A12 Assay is a 384-well multiplex immunoassay kit for the quantitative measurement of S100A12 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The assay requires a small sample volume and has a testing time of approximately 3 5 hours It includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls The kit can perform 38 samples in duplicate along with calibrators and controls