Learn More
Meso Scale Discovery UPLEX HUM S100A12 ASYQP
SDP
Catalog No. 5000081650
Encompass
The Meso Scale Discovery U-PLEX Human S100A12 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of S100A12 and other biomarkers in human serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a typical lower limit of detection of 0 08 ng/mL and an upper limit of 10 000 ng/mL for S100A12
Catalog No. 50-000-81650 Supplier Meso Scale Discovery Supplier No. K151AHPK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human S100A12 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of S100A12 and other biomarkers in human serum plasma and cell culture supernatants The electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals The assay has a typical lower limit of detection of 0 08 ng/mL and an upper limit of 10 000 ng/mL for S100A12