Learn More
Meso Scale Discovery UPLEX HUM SAA ASY384well
SDP
Catalog No. 5000079406
Encompass
The Meso Scale Discovery U-PLEX Human SAA 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for a sensitive reproducible assay with a broad dynamic range The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents It has a sensitivity of 0 3 pg/mL and an assay range of 0 3-2500 pg/mL suitable for high-throughput screening in clinical research
Catalog No. 50-000-79406 Supplier Meso Scale Discovery Supplier No. K251F9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SAA 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying serum amyloid A (SAA) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for a sensitive reproducible assay with a broad dynamic range The kit includes a 384-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents It has a sensitivity of 0 3 pg/mL and an assay range of 0 3-2500 pg/mL suitable for high-throughput screening in clinical research