Learn More
Meso Scale Discovery UPLEX HUM SDF1 ASY 25 PL
SDP
Catalog No. 5000081430
Encompass
The Meso Scale Discovery U-PLEX Human SDF-1alpha Assay Kit is a multiplex immunoassay that measures human SDF-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent It allows for the quantitative measurement of human SDF-1alpha with a lower limit of detection of 1 53 pg/mL and an upper limit of 10 000 pg/mL The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81430 Supplier Meso Scale Discovery Supplier No. K151VBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SDF-1alpha Assay Kit is a multiplex immunoassay that measures human SDF-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and diluent It allows for the quantitative measurement of human SDF-1alpha with a lower limit of detection of 1 53 pg/mL and an upper limit of 10 000 pg/mL The assay takes approximately 3 5 hours to complete