Learn More
Meso Scale Discovery UPLEX HUM SDF1 ASY384well
SDP
Catalog No. 5000079416
Encompass
The Meso Scale Discovery U-PLEX Human SDF-1alpha 384-Well Assay Kit is a multiplex immunoassay that measures SDF-1alpha levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient and high-throughput analysis of up to 80 samples in duplicate It includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator diluent and a vial of calibrator The kit can detect SDF-1alpha concentrations from 0 08-500 pg/mL with an assay time of 3 5 hours
Catalog No. 50-000-79416 Supplier Meso Scale Discovery Supplier No. K251VBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SDF-1alpha 384-Well Assay Kit is a multiplex immunoassay that measures SDF-1alpha levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient and high-throughput analysis of up to 80 samples in duplicate It includes a plate coated with capture antibodies detection antibodies read buffer wash buffer calibrator diluent and a vial of calibrator The kit can detect SDF-1alpha concentrations from 0 08-500 pg/mL with an assay time of 3 5 hours