Learn More
Meso Scale Discovery UPLEX HUM SDF1 ASY384well
SDP
Catalog No. 5000080172
Encompass
The Meso Scale Discovery U-PLEX Human SDF-1alpha 384-Well Assay Kit is a multiplex immunoassay for measuring SDF-1alpha in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies and calibrators Sensitivity is 0 26 pg/mL with a quantification limit of 10 000 pg/mL It requires 25 L of sample and takes about 3 5 hours This aids in studying chemokine biology and inflammation
Catalog No. 50-000-80172 Supplier Meso Scale Discovery Supplier No. K251VBK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SDF-1alpha 384-Well Assay Kit is a multiplex immunoassay for measuring SDF-1alpha in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies and calibrators Sensitivity is 0 26 pg/mL with a quantification limit of 10 000 pg/mL It requires 25 L of sample and takes about 3 5 hours This aids in studying chemokine biology and inflammation