Learn More
Meso Scale Discovery UPLEX HUM Serpin A1 ASY
SDP
Catalog No. 5000082366
Encompass
The Meso Scale Discovery U-PLEX Human Serpin A1 Assay is a multiplex immunoassay kit for measuring Serpin A1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay requires 50 L of sample and is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 The kit includes a plate with capture antibodies detection antibodies and necessary calibrators and controls Turnaround time is 3 5 hours
Catalog No. 50-000-82366 Supplier Meso Scale Discovery Supplier No. K151W9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Serpin A1 Assay is a multiplex immunoassay kit for measuring Serpin A1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The assay requires 50 L of sample and is compatible with Meso Scale Discovery instruments like the MESO QuickPlex SQ 120 The kit includes a plate with capture antibodies detection antibodies and necessary calibrators and controls Turnaround time is 3 5 hours