Learn More
Meso Scale Discovery UPLEX HUM Serpin A1 ASYQP
SDP
Catalog No. 5000081961
Encompass
The Meso Scale Discovery U-PLEX Human Serpin A1 Assay Kit is designed to quantitatively measure Serpin A1 levels in human serum plasma and cell culture supernatants This electrochemiluminescent multiplex immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate detection of Serpin A1 on the MESO QuickPlex SQ 120 instrument The assay has a detection range of 50-10 000 pg/mL and includes a calibrator and control to ensure reliable results This kit offers a simple rapid way to analyze Serpin A1 an important biomarker from small sample volumes
Catalog No. 50-000-81961 Supplier Meso Scale Discovery Supplier No. K151W9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Serpin A1 Assay Kit is designed to quantitatively measure Serpin A1 levels in human serum plasma and cell culture supernatants This electrochemiluminescent multiplex immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate detection of Serpin A1 on the MESO QuickPlex SQ 120 instrument The assay has a detection range of 50-10 000 pg/mL and includes a calibrator and control to ensure reliable results This kit offers a simple rapid way to analyze Serpin A1 an important biomarker from small sample volumes