Learn More
Meso Scale Discovery UPLEX HUM SHBG ASY384well
SDP
Catalog No. 5000079480
Encompass
The Meso Scale Discovery U-PLEX Human SHBG 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of sex hormone-binding globulin (SHBG) in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a rapid ultra-sensitive immunoassay with an extended dynamic range The kit includes a 384-well plate coated with an SHBG capture antibody detection antibody conjugated to an MSD SULFO-TAG calibrator and sample diluent The assay has a typical sensitivity of 0 05 nmol/L and an upper limit of quantification of 200 nmol/L
Catalog No. 50-000-79480 Supplier Meso Scale Discovery Supplier No. K251G9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SHBG 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of sex hormone-binding globulin (SHBG) in human serum plasma and cell culture supernatants The assay uses MSD GOLD SULFO-TAG labels and MSD high bind plates to provide a rapid ultra-sensitive immunoassay with an extended dynamic range The kit includes a 384-well plate coated with an SHBG capture antibody detection antibody conjugated to an MSD SULFO-TAG calibrator and sample diluent The assay has a typical sensitivity of 0 05 nmol/L and an upper limit of quantification of 200 nmol/L