Learn More
Meso Scale Discovery UPLEX HUM SHBG ASYQP 25 PL
SDP
Catalog No. 5000082012
Encompass
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit is a multiplex immunoassay that measures human sex hormone-binding globulin (SHBG) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity and precision with minimal cross-reactivity It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators controls 10X read buffer sample diluent wash buffer and plate seals The kit has a 4 5 hour total assay time allowing rapid processing of large sample sets
Catalog No. 50-000-82012 Supplier Meso Scale Discovery Supplier No. K151G9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human SHBG Assay Kit is a multiplex immunoassay that measures human sex hormone-binding globulin (SHBG) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity and precision with minimal cross-reactivity It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators controls 10X read buffer sample diluent wash buffer and plate seals The kit has a 4 5 hour total assay time allowing rapid processing of large sample sets