Learn More
Meso Scale Discovery UPLEX HUM sTfR1 ASY 25 PL
SDP
Catalog No. 5000080584
Encompass
The Meso Scale Discovery U-PLEX Human STFR-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of soluble transferrin receptor 1 (sTFR1) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the rapid and efficient analysis of sTFR1 an important biomarker of iron status and erythropoiesis With minimal hands-on time this kit serves as a useful tool for research and drug development applications involving iron metabolism and red blood cell production
Catalog No. 50-000-80584 Supplier Meso Scale Discovery Supplier No. K151P9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human STFR-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of soluble transferrin receptor 1 (sTFR1) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay allows for the rapid and efficient analysis of sTFR1 an important biomarker of iron status and erythropoiesis With minimal hands-on time this kit serves as a useful tool for research and drug development applications involving iron metabolism and red blood cell production