Learn More
Meso Scale Discovery UPLEX HUM sTfR1 ASY384well
SDP
Catalog No. 5000079405
Encompass
The U-PLEX Human STFR 1-384 Well Assay from Meso Scale Diagnostics is a 384-well multiplex immunoassay that measures soluble ST2 free T3 free T4 and reverse T3 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection for high sensitivity and broad dynamic range The assay requires 10 L of sample per well and the kit provides reagents for testing 384 samples across 4 plates Calibration curves quality controls reagents and 384-well plates are included for ease-of-use This quantitative immunoassay allows high-throughput analysis of ST2 and thyroid hormone levels
Catalog No. 50-000-79405 Supplier Meso Scale Discovery Supplier No. K251P9K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human STFR 1-384 Well Assay from Meso Scale Diagnostics is a 384-well multiplex immunoassay that measures soluble ST2 free T3 free T4 and reverse T3 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection for high sensitivity and broad dynamic range The assay requires 10 L of sample per well and the kit provides reagents for testing 384 samples across 4 plates Calibration curves quality controls reagents and 384-well plates are included for ease-of-use This quantitative immunoassay allows high-throughput analysis of ST2 and thyroid hormone levels