Learn More
Meso Scale Discovery UPLEX HUM sTfR1 ASYQP 25 PL
SDP
Catalog No. 5000080583
Encompass
The U-PLEX Human STFR-1 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures soluble transferrin receptor 1 (sTfR1) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the quantitative measurement of sTfR1 which can aid in the assessment of iron status and erythropoiesis The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control
Catalog No. 50-000-80583 Supplier Meso Scale Discovery Supplier No. K151P9K24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human STFR-1 Assay from Meso Scale Diagnostics is a multiplex immunoassay that measures soluble transferrin receptor 1 (sTfR1) in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and requires low sample volume This assay allows for the quantitative measurement of sTfR1 which can aid in the assessment of iron status and erythropoiesis The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer calibrator and control