Learn More
Meso Scale Discovery UPLEX HUM TGFB1 ASY 25 PL
SDP
Catalog No. 5000078929
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta1 Assay Kit is designed for the quantitative measurement of active and total TGF-beta1 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and buffers for sample dilution and washing The assay has a typical sensitivity of 0 3 pg/mL and range of 0 3-2000 pg/mL Sample incubation time is 3 hours
Catalog No. 50-000-78929 Supplier Meso Scale Discovery Supplier No. K151XWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TGF-beta1 Assay Kit is designed for the quantitative measurement of active and total TGF-beta1 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and buffers for sample dilution and washing The assay has a typical sensitivity of 0 3 pg/mL and range of 0 3-2000 pg/mL Sample incubation time is 3 hours