Learn More
Meso Scale Discovery UPLEX HUM TGFB1 ASY384well
SDP
Catalog No. 5000079401
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta1 384-well Assay Kit is a multiplex immunoassay that measures TGF-beta1 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for high throughput screening and only requires 10-25 L sample volume per well The kit includes a 384-well MULTI-ARRAY Plate pre-coated with capture antibodies detection antibodies read buffer calibrators and controls needed to quantify TGF-beta1 levels It has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79401 Supplier Meso Scale Discovery Supplier No. K251XWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TGF-beta1 384-well Assay Kit is a multiplex immunoassay that measures TGF-beta1 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for high throughput screening and only requires 10-25 L sample volume per well The kit includes a 384-well MULTI-ARRAY Plate pre-coated with capture antibodies detection antibodies read buffer calibrators and controls needed to quantify TGF-beta1 levels It has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL