Learn More
Meso Scale Discovery UPLEX HUM TGFB1 ASY384well
SDP
Catalog No. 5000080519
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta1 384-Well Assay Kit is a multiplex immunoassay that measures active and total TGF-beta1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a 384-well plate format The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and Calibrator It can detect TGF-beta1 levels between 1 7 pg/mL to 700 pg/mL with an assay time of 3 5 hours
Catalog No. 50-000-80519 Supplier Meso Scale Discovery Supplier No. K251XWK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TGF-beta1 384-Well Assay Kit is a multiplex immunoassay that measures active and total TGF-beta1 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a 384-well plate format The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Sample Diluent and Calibrator It can detect TGF-beta1 levels between 1 7 pg/mL to 700 pg/mL with an assay time of 3 5 hours