Learn More
Meso Scale Discovery UPLEX HUM TGFB2 ASY384well
SDP
Catalog No. 5000079476
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta2 384-Well Assay Kit is a multiplex immunoassay that measures human TGF-beta2 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a 384-well plate format This assay requires a Meso Scale Discovery SECTOR Imager and provides reagents for one 384-well plate to perform up to 380 tests in duplicate
Catalog No. 50-000-79476 Supplier Meso Scale Discovery Supplier No. K251XUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TGF-beta2 384-Well Assay Kit is a multiplex immunoassay that measures human TGF-beta2 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a 384-well plate format This assay requires a Meso Scale Discovery SECTOR Imager and provides reagents for one 384-well plate to perform up to 380 tests in duplicate