Learn More
Meso Scale Discovery UPLEX HUM TGFB3 ASY 25 PL
SDP
Catalog No. 5000080107
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta3 Assay Kit measures TGF-beta3 in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification down to sub-pg/mL levels It includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent The assay runs in under 5 hours with a 50 L sample offering high sensitivity and minimal matrix interference for studying TGF-beta3 biology and therapies
Catalog No. 50-000-80107 Supplier Meso Scale Discovery Supplier No. K151XVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TGF-beta3 Assay Kit measures TGF-beta3 in human serum plasma and cell culture supernatants This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive quantification down to sub-pg/mL levels It includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and diluent The assay runs in under 5 hours with a 50 L sample offering high sensitivity and minimal matrix interference for studying TGF-beta3 biology and therapies