Learn More
Meso Scale Discovery UPLEX HUM TGFB3 ASY384well
SDP
Catalog No. 5000079477
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta3 384-Well Assay Kit is a multiplex immunoassay that measures TGF-beta3 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient and high-throughput analysis of up to 80 samples in duplicate It includes a plate coated with capture antibodies detection antibodies Read Buffer and calibrator This quantitative assay takes approximately 3 5 hours to complete and has a lower limit of detection of 1 7 pg/mL for TGF-beta3
Catalog No. 50-000-79477 Supplier Meso Scale Discovery Supplier No. K251XVK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TGF-beta3 384-Well Assay Kit is a multiplex immunoassay that measures TGF-beta3 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for efficient and high-throughput analysis of up to 80 samples in duplicate It includes a plate coated with capture antibodies detection antibodies Read Buffer and calibrator This quantitative assay takes approximately 3 5 hours to complete and has a lower limit of detection of 1 7 pg/mL for TGF-beta3