Learn More
Meso Scale Discovery UPLEX HUM TGFB3 ASY384well
SDP
Catalog No. 5000079856
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta3 384-Well Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of the human cytokine TGF-beta3 This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The assay has a detection range of 1 5-2500 pg/mL and includes reagents for standard curve generation This quantitative immunoassay provides sensitive accurate and reproducible measurement of TGF-beta3 levels in human serum plasma cell culture supernatants and other sample types
Catalog No. 50-000-79856 Supplier Meso Scale Discovery Supplier No. K251XVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TGF-beta3 384-Well Assay is a multiplex immunoassay kit that enables simultaneous detection and quantification of the human cytokine TGF-beta3 This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The assay has a detection range of 1 5-2500 pg/mL and includes reagents for standard curve generation This quantitative immunoassay provides sensitive accurate and reproducible measurement of TGF-beta3 levels in human serum plasma cell culture supernatants and other sample types