Learn More
Meso Scale Discovery UPLEX HUM TGFB3 ASYQP 25 PL
SDP
Catalog No. 5000079670
Encompass
The Meso Scale Discovery U-PLEX Human TGF-beta3 Assay Kit is designed for the quantitative measurement of active and total TGF-beta3 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity and a broad dynamic range This multiplex kit includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and quantify sample concentrations The assay has high specificity for active and total TGF-beta3 with minimal cross-reactivity to related analytes
Catalog No. 50-000-79670 Supplier Meso Scale Discovery Supplier No. K151XVK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TGF-beta3 Assay Kit is designed for the quantitative measurement of active and total TGF-beta3 in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses MSD s proprietary QuickPlex SQ 120 instrument and provides high sensitivity and a broad dynamic range This multiplex kit includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and quantify sample concentrations The assay has high specificity for active and total TGF-beta3 with minimal cross-reactivity to related analytes