Learn More
Meso Scale Discovery UPLEX HUM Tie2 ASY384well
SDP
Catalog No. 5000079857
Encompass
The Meso Scale Discovery U-PLEX Human TIE-2 384-Well Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions It allows simultaneous measurement of total TIE-2 levels in 384 samples in just 3 5 hours
Catalog No. 50-000-79857 Supplier Meso Scale Discovery Supplier No. K251F8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TIE-2 384-Well Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions It allows simultaneous measurement of total TIE-2 levels in 384 samples in just 3 5 hours