Learn More
Meso Scale Discovery UPLEX HUM Tie2 ASYQP 25 PL
SDP
Catalog No. 5000082097
Encompass
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Wash Buffer Diluent 100 and calibrators and controls to generate a standard curve and validate assay performance With a measuring time of 2 5 hours this kit allows quantification of human TIE-2 levels with accuracy and precision
Catalog No. 50-000-82097 Supplier Meso Scale Discovery Supplier No. K151F8K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TIE-2 Assay Kit is a multiplex immunoassay that measures human TIE-2 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Wash Buffer Diluent 100 and calibrators and controls to generate a standard curve and validate assay performance With a measuring time of 2 5 hours this kit allows quantification of human TIE-2 levels with accuracy and precision