Learn More
Meso Scale Discovery UPLEX HUM TIGIT ASYQP 25 PL
SDP
Catalog No. 5000081733
Encompass
The Meso Scale Discovery U-PLEX Human TIGIT Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TIGIT and other biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve It allows efficient and reproducible quantification of TIGIT levels relevant to immunology and oncology research
Catalog No. 50-000-81733 Supplier Meso Scale Discovery Supplier No. K151W7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TIGIT Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TIGIT and other biomarkers in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators to generate a standard curve It allows efficient and reproducible quantification of TIGIT levels relevant to immunology and oncology research