Learn More
Meso Scale Discovery UPLEX HUM TLR1 ASYQP 25 PL
SDP
Catalog No. 5000079952
Encompass
The U-PLEX Human TLR1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Toll-like receptor 1 (TLR1) in cell and tissue lysates This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and reagents needed to perform the assay Results can be read on Meso Scale Discovery instrumentation such as the QuickPlex SQ 120 This assay allows for the quantitative measurement of TLR1 which plays a key role in innate immunity
Catalog No. 50-000-79952 Supplier Meso Scale Discovery Supplier No. K151J7K24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human TLR1 Assay Kit from Meso Scale Discovery is a multiplex immunoassay that measures human Toll-like receptor 1 (TLR1) in cell and tissue lysates This assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and reagents needed to perform the assay Results can be read on Meso Scale Discovery instrumentation such as the QuickPlex SQ 120 This assay allows for the quantitative measurement of TLR1 which plays a key role in innate immunity