Learn More
Meso Scale Discovery UPLEX HUM TNFB ASY384well
SDP
Catalog No. 5000080178
Encompass
The Meso Scale Discovery U-PLEX Human TNF-beta 384-Well Assay Kit is a multiplex immunoassay that measures TNF-beta levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls It allows simultaneous measurement of TNF-beta and other biomarkers in a small sample volume The assay takes approximately 3 hours to complete
Catalog No. 50-000-80178 Supplier Meso Scale Discovery Supplier No. K251UUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TNF-beta 384-Well Assay Kit is a multiplex immunoassay that measures TNF-beta levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls It allows simultaneous measurement of TNF-beta and other biomarkers in a small sample volume The assay takes approximately 3 hours to complete