Learn More
Meso Scale Discovery UPLEX HUM TNFRI ASY 25 PL
SDP
Catalog No. 5000081660
Encompass
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of TNF RI in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance It allows rapid and accurate quantification of TNF RI levels relevant to inflammation and autoimmune disease research
Catalog No. 50-000-81660 Supplier Meso Scale Discovery Supplier No. K151AHQK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of TNF RI in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance It allows rapid and accurate quantification of TNF RI levels relevant to inflammation and autoimmune disease research