Learn More
Meso Scale Discovery UPLEX HUM TNFRI ASY384well
SDP
Catalog No. 5000080816
Encompass
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a 384-well multiplex immunoassay kit for quantifying human TNF RI in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision The assay measures TNF RI levels from 0 03 to 1000 pg/mL with a 25 L sample per well The kit includes a 384-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and controls allowing simultaneous measurement of TNF RI and other biomarkers on the U-PLEX platform
Catalog No. 50-000-80816 Supplier Meso Scale Discovery Supplier No. K251AHQK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a 384-well multiplex immunoassay kit for quantifying human TNF RI in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision The assay measures TNF RI levels from 0 03 to 1000 pg/mL with a 25 L sample per well The kit includes a 384-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and controls allowing simultaneous measurement of TNF RI and other biomarkers on the U-PLEX platform