Learn More
Meso Scale Discovery UPLEX HUM TNFRI ASY384well
SDP
Catalog No. 5000081122
Encompass
The Meso Scale Discovery U-PLEX Human TNF RI 384-Well Assay Kit is designed for the quantitative measurement of human TNF RI in serum plasma cell culture supernatants cell and tissue lysates It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid time to results in a 384-well plate format The assay requires a low sample volume of 25 L and has a typical lower limit of detection of 19 pg/mL The kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator Diluent and vials of lyophilized Calibrator
Catalog No. 50-000-81122 Supplier Meso Scale Discovery Supplier No. K251AHQK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TNF RI 384-Well Assay Kit is designed for the quantitative measurement of human TNF RI in serum plasma cell culture supernatants cell and tissue lysates It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid time to results in a 384-well plate format The assay requires a low sample volume of 25 L and has a typical lower limit of detection of 19 pg/mL The kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator Diluent and vials of lyophilized Calibrator