Learn More
Meso Scale Discovery UPLEX HUM TNFRI ASYQP 25 PL
SDP
Catalog No. 5000079092
Encompass
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TNF receptor I (TNFR1) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay It can detect TNFR1 levels as low as 17 pg/mL with an upper limit of 100 000 pg/mL The total assay time is approximately 3 5 hours
Catalog No. 50-000-79092 Supplier Meso Scale Discovery Supplier No. K151AHQK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TNF RI Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human TNF receptor I (TNFR1) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls needed to perform the assay It can detect TNFR1 levels as low as 17 pg/mL with an upper limit of 100 000 pg/mL The total assay time is approximately 3 5 hours