Learn More
Meso Scale Discovery UPLEX HUM TNFRII ASY 25 PL
SDP
Catalog No. 5000079560
Encompass
The Meso Scale Discovery U-PLEX Human TNF RII Assay is a multiplex immunoassay kit that enables simultaneous quantification of human TNF receptor II (TNFRII) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals sufficient for one 96-well plate It can accurately measure human TNFRII concentrations from 50-100 000 pg/mL with minimal cross-reactivity
Catalog No. 50-000-79560 Supplier Meso Scale Discovery Supplier No. K151AHRK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TNF RII Assay is a multiplex immunoassay kit that enables simultaneous quantification of human TNF receptor II (TNFRII) in serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X read buffer 10X wash buffer and plate seals sufficient for one 96-well plate It can accurately measure human TNFRII concentrations from 50-100 000 pg/mL with minimal cross-reactivity