Learn More
Meso Scale Discovery UPLEX HUM TNFRII ASY384well
SDP
Catalog No. 5000080275
Encompass
The Meso Scale Discovery TNF-RII Singleplex Assay Kit is a highly sensitive immunoassay that quantitatively measures soluble human TNF-RII in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) technology on patterned arrays for high performance and utilizes MSD s patented SULFO-TAG labels for ultra-high sensitivity with no wash steps The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and diluent The assay has a typical lower limit of detection of 3 5 pg/mL and an upper limit of 10 000 pg/mL This product enables quantification of TNF-RII for research use only
Catalog No. 50-000-80275 Supplier Meso Scale Discovery Supplier No. K251AHRK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery TNF-RII Singleplex Assay Kit is a highly sensitive immunoassay that quantitatively measures soluble human TNF-RII in serum plasma and cell culture supernatants This assay uses electrochemiluminescence (ECL) technology on patterned arrays for high performance and utilizes MSD s patented SULFO-TAG labels for ultra-high sensitivity with no wash steps The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and diluent The assay has a typical lower limit of detection of 3 5 pg/mL and an upper limit of 10 000 pg/mL This product enables quantification of TNF-RII for research use only