Learn More
Meso Scale Discovery UPLEX HUM TNFRII ASY384well
SDP
Catalog No. 5000081001
Encompass
The Meso Scale Discovery TNF-RII Kit is a highly sensitive assay for measuring soluble tumor necrosis factor receptor II (sTNF-RII) in serum plasma and cell culture supernatants The kit uses Meso Scale Discovery s electrochemiluminescent detection technology and provides accurate reproducible results in just 3 5 hours The assay has a lower limit of detection of 3 5 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent and a vial of lyophilized calibrator The TNF-RII Kit allows for the precise sensitive and rapid quantification of sTNF-RII from a variety of sample types
Catalog No. 50-000-81001 Supplier Meso Scale Discovery Supplier No. K251AHRK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery TNF-RII Kit is a highly sensitive assay for measuring soluble tumor necrosis factor receptor II (sTNF-RII) in serum plasma and cell culture supernatants The kit uses Meso Scale Discovery s electrochemiluminescent detection technology and provides accurate reproducible results in just 3 5 hours The assay has a lower limit of detection of 3 5 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent and a vial of lyophilized calibrator The TNF-RII Kit allows for the precise sensitive and rapid quantification of sTNF-RII from a variety of sample types