Learn More
Meso Scale Discovery UPLEX HUM TPO ASY384well
SDP
Catalog No. 5000080663
Encompass
The Meso Scale Discovery U-PLEX Human TPO 384-Well Assay Kit is a multiplex immunoassay that measures total TPO in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has a typical sensitivity of 0 08 pg/mL and an assay range from 0 156 pg/mL to 2500 pg/mL It requires 2 5 L sample size and the total assay time is approximately 3 5 hours This assay kit includes a 384-well standard microplate detection antibody read buffer 10X wash buffer blocker A calibration curve fit software and an assay protocol
Catalog No. 50-000-80663 Supplier Meso Scale Discovery Supplier No. K251VKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TPO 384-Well Assay Kit is a multiplex immunoassay that measures total TPO in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay allowing high throughput screening The assay has a typical sensitivity of 0 08 pg/mL and an assay range from 0 156 pg/mL to 2500 pg/mL It requires 2 5 L sample size and the total assay time is approximately 3 5 hours This assay kit includes a 384-well standard microplate detection antibody read buffer 10X wash buffer blocker A calibration curve fit software and an assay protocol