Learn More
Meso Scale Discovery UPLEX HUM TRAIL ASY384well
SDP
Catalog No. 5000080682
Encompass
The U-PLEX Human TRAIL 384-Well Assay Kit from Meso Scale Discovery measures TRAIL in human serum plasma and cell culture supernatants using electrochemiluminescent detection It offers high sensitivity and a broad dynamic range with a detection limit of 0 04 pg/mL and quantification up to 2500 pg/mL The kit includes a 384-well plate antibodies Read Buffer Calibrator Diluent and U-PLEX Calibrator Blend allowing up to 380 tests in about 3 5 hours This quantitative immunoassay streamlines TRAIL measurement
Catalog No. 50-000-80682 Supplier Meso Scale Discovery Supplier No. K251XTK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human TRAIL 384-Well Assay Kit from Meso Scale Discovery measures TRAIL in human serum plasma and cell culture supernatants using electrochemiluminescent detection It offers high sensitivity and a broad dynamic range with a detection limit of 0 04 pg/mL and quantification up to 2500 pg/mL The kit includes a 384-well plate antibodies Read Buffer Calibrator Diluent and U-PLEX Calibrator Blend allowing up to 380 tests in about 3 5 hours This quantitative immunoassay streamlines TRAIL measurement