Learn More
Meso Scale Discovery UPLEX HUM TSLP ASY384well
SDP
Catalog No. 5000079855
Encompass
The Meso Scale Discovery U-PLEX Human TSLP 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human thymic stromal lymphopoietin (TSLP) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined workflow This multiplex kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls needed to quantify TSLP levels The kit is compatible with the MSD QuickPlex SQ 120 MESO QuickPlex SQ 120 and other MSD instruments
Catalog No. 50-000-79855 Supplier Meso Scale Discovery Supplier No. K251WHK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human TSLP 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human thymic stromal lymphopoietin (TSLP) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a streamlined workflow This multiplex kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls needed to quantify TSLP levels The kit is compatible with the MSD QuickPlex SQ 120 MESO QuickPlex SQ 120 and other MSD instruments