Learn More
Meso Scale Discovery UPLEX HUM TSLP ASYQP 25 PL
SDP
Catalog No. 5000079702
Encompass
The U-PLEX Human TSLP Assay (K151WHK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of thymic stromal lymphopoietin (TSLP) in human biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable TSLP quantification in research applications
Catalog No. 50-000-79702 Supplier Meso Scale Discovery Supplier No. K151WHK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human TSLP Assay (K151WHK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of thymic stromal lymphopoietin (TSLP) in human biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable TSLP quantification in research applications