Learn More
Meso Scale Discovery UPLEX HUM VCAM1 ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151H9K4
Encompass
The Meso Scale Discovery U-PLEX Human VCAM-1 Assay Kit quantifies VCAM-1 in human serum plasma and cell culture supernatants using electrochemiluminescence It employs MSD GOLD Streptavidin Multi-Array 96-well plates and a VCAM-1 capture antibody A SULFO-TAG labeled detection antibody identifies the analyte Upon applying electricity the SULFO-TAG emits light measured by an MSD instrument The kit includes a VCAM-1 calibrator and control for standard curve generation and assay validation detecting VCAM-1 from 0 05-400 ng/mL
Catalog No. 50-000-80792
Add to Cart
Request a Quote