Learn More
Meso Scale Discovery UPLEX HUM VCAM1 ASY384well
SDP
Catalog No. 5000080567
Encompass
The Meso Scale Discovery U-PLEX Human VCAM-1 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of VCAM-1 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput screening and includes VCAM-1 calibrators and positive controls It can be used for biomarker research immunology studies and drug discovery and development
Catalog No. 50-000-80567 Supplier Meso Scale Discovery Supplier No. K251H9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VCAM-1 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of VCAM-1 in human serum plasma and cell culture supernatants It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to a 7-log dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput screening and includes VCAM-1 calibrators and positive controls It can be used for biomarker research immunology studies and drug discovery and development