Learn More
Meso Scale Discovery UPLEX HUM VCAM1 ASY384well
SDP
Catalog No. 5000080657
Encompass
The Meso Scale Discovery U-PLEX Human VCAM-1 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of VCAM-1 in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput analysis of VCAM-1 an important biomarker of endothelial dysfunction and cardiovascular disease risk With a rapid 2 5 hour protocol this assay kit offers an efficient way to quantitatively analyze VCAM-1 levels
Catalog No. 50-000-80657 Supplier Meso Scale Discovery Supplier No. K251H9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VCAM-1 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of VCAM-1 in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This 384-well plate assay allows high-throughput analysis of VCAM-1 an important biomarker of endothelial dysfunction and cardiovascular disease risk With a rapid 2 5 hour protocol this assay kit offers an efficient way to quantitatively analyze VCAM-1 levels