Learn More
Meso Scale Discovery UPLEX HUM VEGFa ASY 25 PL
SDP
Catalog No. 5000080372
Encompass
The Meso Scale Discovery U-PLEX Human VEGF-A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of vascular endothelial growth factor A (VEGF-A) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 15 6-2000 pg/mL and shows no cross-reactivity with other human VEGF isoforms The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators Sample incubation time is just 2 hours The assay is for research use only
Catalog No. 50-000-80372 Supplier Meso Scale Discovery Supplier No. K151UVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VEGF-A Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of vascular endothelial growth factor A (VEGF-A) in human serum plasma and cell culture supernatants It utilizes MSD s proprietary U-PLEX technology is highly sensitive has a broad dynamic range of 15 6-2000 pg/mL and shows no cross-reactivity with other human VEGF isoforms The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators Sample incubation time is just 2 hours The assay is for research use only