Learn More
Meso Scale Discovery UPLEX HUM VEGFA ASY384well
SDP
Catalog No. 5000080179
Encompass
The Meso Scale Discovery U-PLEX Human VEGF-A 384-Well Assay Kit is a multiplex immunoassay for measuring VEGF-A in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 384-well plate with capture antibodies detection antibodies and calibrators It allows simultaneous measurement of VEGF-A and other biomarkers on the U-PLEX platform aiding in the study of angiogenesis Designed for the MESO QuickPlex SQ 120 instrument it offers high performance
Catalog No. 50-000-80179 Supplier Meso Scale Discovery Supplier No. K251UVK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VEGF-A 384-Well Assay Kit is a multiplex immunoassay for measuring VEGF-A in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a 384-well plate with capture antibodies detection antibodies and calibrators It allows simultaneous measurement of VEGF-A and other biomarkers on the U-PLEX platform aiding in the study of angiogenesis Designed for the MESO QuickPlex SQ 120 instrument it offers high performance