Learn More
Meso Scale Discovery UPLEX HUM VEGFD ASY 25 PL
SDP
Catalog No. 5000082030
Encompass
The Meso Scale Discovery U-PLEX Human VEGF-D Assay Kit is a multiplex immunoassay for measuring VEGF-D in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity up to 5 logs of dynamic range and low sample volume needs The assay quantifies VEGF-D as low as 0 06 pg/mL with 25 L of sample It includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals The kit is highly specific with no cross-reactivity enabling rapid and accurate VEGF-D quantification from various samples
Catalog No. 50-000-82030 Supplier Meso Scale Discovery Supplier No. K151C8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VEGF-D Assay Kit is a multiplex immunoassay for measuring VEGF-D in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity up to 5 logs of dynamic range and low sample volume needs The assay quantifies VEGF-D as low as 0 06 pg/mL with 25 L of sample It includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals The kit is highly specific with no cross-reactivity enabling rapid and accurate VEGF-D quantification from various samples