Learn More
Meso Scale Discovery UPLEX HUM VEGFD ASY384well
SDP
Catalog No. 5000078663
Encompass
The Meso Scale Discovery Human VEGF-D Singleplex Assay Kit is a singleplex immunoassay that allows for the quantitative measurement of human VEGF-D in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay can detect VEGF-D concentrations as low as 3 5 pg/mL with an upper limit of 10 000 pg/mL It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators The total assay time is 3 5 hours
Catalog No. 50-000-78663 Supplier Meso Scale Discovery Supplier No. K251C8K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Human VEGF-D Singleplex Assay Kit is a singleplex immunoassay that allows for the quantitative measurement of human VEGF-D in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay can detect VEGF-D concentrations as low as 3 5 pg/mL with an upper limit of 10 000 pg/mL It includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators The total assay time is 3 5 hours