Learn More
Meso Scale Discovery UPLEX HUM VEGFD ASY384well
SDP
Catalog No. 5000080279
Encompass
The Meso Scale Discovery VEGF-D Kit is an immunoassay that quantitatively measures human VEGF-D in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay can detect VEGF-D concentrations as low as 3 pg/mL and has a 7-log dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator diluent and a calibrator The total assay time is 3 5 hours
Catalog No. 50-000-80279 Supplier Meso Scale Discovery Supplier No. K251C8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery VEGF-D Kit is an immunoassay that quantitatively measures human VEGF-D in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay can detect VEGF-D concentrations as low as 3 pg/mL and has a 7-log dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker solutions calibrator diluent and a calibrator The total assay time is 3 5 hours