Learn More
Meso Scale Discovery UPLEX HUM VEGFR1/Flt1 ASY
SDP
Catalog No. 5000081580
Encompass
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a multiplex immunoassay kit that enables the simultaneous measurement of VEGF Receptor 1 (VEGFR-1) and other biomarkers in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology providing high sensitivity a wide dynamic range and compatibility with small sample volumes The assay requires an MSD instrument such as QuickPlex SQ 120 or HD-1 for detection With a rapid 2 5 hour protocol this assay kit offers a streamlined workflow for quantifying VEGFR-1 levels alongside other biomarkers
Catalog No. 50-000-81580 Supplier Meso Scale Discovery Supplier No. K151AHHK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a multiplex immunoassay kit that enables the simultaneous measurement of VEGF Receptor 1 (VEGFR-1) and other biomarkers in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR technology providing high sensitivity a wide dynamic range and compatibility with small sample volumes The assay requires an MSD instrument such as QuickPlex SQ 120 or HD-1 for detection With a rapid 2 5 hour protocol this assay kit offers a streamlined workflow for quantifying VEGFR-1 levels alongside other biomarkers