Learn More
Meso Scale Discovery UPLEX HUM VEGFR1/Flt1 ASYQP
SDP
Catalog No. 5000079087
Encompass
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a multiplex immunoassay kit for measuring human VEGFR-1 and other biomarkers in serum plasma and cell culture supernatants It uses MSD s SECTOR technology for high sensitivity and a large dynamic range with small sample volumes The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals It quantifies VEGFR-1 levels with a detection limit of 0 07 ng/mL
Catalog No. 50-000-79087 Supplier Meso Scale Discovery Supplier No. K151AHHK22
The Meso Scale Discovery U-PLEX Human VEGFR-1 (FLT-1) Assay is a multiplex immunoassay kit for measuring human VEGFR-1 and other biomarkers in serum plasma and cell culture supernatants It uses MSD s SECTOR technology for high sensitivity and a large dynamic range with small sample volumes The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals It quantifies VEGFR-1 levels with a detection limit of 0 07 ng/mL